Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 9 | 2449 | ||
XM_005269491.1 | XP_005269548.1 | 9 | 2519 | |||
XM_005269492.2 | XP_005269549.1 | 7 | 1812 | |||
XM_011539216.1 | XP_011537518.1 | 10 | 3177 | |||
XM_011539217.2 | XP_011537519.1 | 10 | 2872 | |||
XM_011539220.2 | XP_011537522.1 | 10 | 2417 | |||
XM_011539222.2 | XP_011537524.1 | 10 | 2539 | |||
XM_011539223.2 | XP_011537525.1 | 9 | 3227 | |||
XM_011539224.2 | XP_011537526.1 | 9 | 2373 | |||
XM_011539226.1 | XP_011537528.1 | 9 | 2372 | |||
XM_011539227.2 | XP_011537529.1 | 9 | 2705 | |||
XM_011539230.2 | XP_011537532.1 | 8 | 2494 | |||
XM_011539231.1 | XP_011537533.1 | 9 | 4504 | |||
XM_011539232.1 | XP_011537534.1 | 9 | 2171 | |||
XM_011539233.1 | XP_011537535.1 | 8 | 1934 | |||
XM_017015634.1 | XP_016871123.1 | 9 | 2886 | |||
GenBank mRNA | AF435960.1 | - | 6 | 956 | ||
AK297577.1 | - | 10 | 2650 | |||
BC021140.1 | - | 6 | 962 |