Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009646.1 | NP_001009646.1 | 2 | 222 | ||
XM_006230246.2 | XP_006230308.1 | 2 | 393 | |||
XM_008759877.1 | XP_008758099.1 | 2 | 698 | |||
XM_008759878.1 | XP_008758100.1 | 3 | 485 | |||
GenBank mRNA | BC088177.1 | - | 2 | 222 | 7323872 | |
FQ209866.1 | - | 2 | 216 |