Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSPRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317943.1 | NP_001304872.1 | 4 | 608 | ||
NM_001317944.1 | NP_001304873.1 | 4 | 593 | |||
NM_017688.2 | NP_060158.2 | 4 | 593 | |||
XM_006717149.3 | XP_006717212.1 | 3, 4 | 581 | |||
GenBank mRNA | AJ276691.1 | - | 4 | 574 | ||
AK000157.1 | - | 4 | 319 | |||
AK092607.1 | - | 4 | 584 | |||
BC001477.2 | - | 4 | 602 | |||
BP370567.1 | - | 4 | 431 | |||
BX462607.2 | - | 4 | 592 |