Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr1275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011795.1 | NP_001011795.1 | 1 | 117 | ||
GenBank mRNA | BC147155.1 | - | 1 | 163 | 9056178 | |
BC147156.1 | - | 1 | 163 | 9056164 |