Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011917.1 | NP_001011917.1 | 10 | 1379 | ||
XM_006252098.2 | XP_006252160.1 | 11 | 1483 | |||
XM_006252099.2 | XP_006252161.1 | 12 | 1567 | |||
XM_006252100.2 | XP_006252162.1 | 13 | 1763 | |||
XM_006252101.2 | XP_006252163.1 | 12 | 1679 | |||
XM_008770742.1 | XP_008768964.1 | 12 | 1576 | |||
XM_008770743.1 | XP_008768965.1 | 9 | 1973 | |||
GenBank mRNA | BC083684.1 | - | 10 | 1379 | 7188949 |