Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011933.1 | NP_001011933.1 | 12 | 1623 | ||
XM_006230017.2 | XP_006230079.1 | 13 | 1705 | |||
XM_006230018.2 | XP_006230080.1 | 13 | 1630 | |||
XM_006230020.2 | XP_006230082.1 | 13 | 1707 | |||
GenBank mRNA | BC082015.1 | - | 12 | 1623 | 7132096 | |
FQ221385.1 | - | 13 | 1689 |