Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011995.1 | NP_001011995.1 | 4 | 1521 | ||
GenBank mRNA | BC078829.1 | - | 4 | 1521 | 7112420 | |
FQ225183.1 | - | 4 | 1542 | |||
FQ226218.1 | - | 4 | 1542 | |||
FQ235315.1 | - | 4 | 1545 |
Gene Symbol: LOC100909441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749354.3 | XP_003749402.1 | 4 | 1532 |