Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013057.1 | NP_001013075.1 | 8 | 922 | ||
XM_006254183.2 | XP_006254245.1 | 8 | 934 | |||
XM_006254184.2 | XP_006254246.1 | 8 | 913 | |||
XM_006254185.2 | XP_006254247.1 | 7 | 856 | |||
GenBank mRNA | BC078957.1 | - | 8 | 922 | 7110806 |