Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013071.1 | NP_001013089.1 | 10 | 1387 | ||
GenBank mRNA | BC091237.1 | - | 10 | 1387 | 7323367 |