Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013104.1 | NP_001013122.1 | 8 | 1945 | ||
XM_008765549.1 | XP_008763771.1 | 9 | 4066 | |||
XM_008765550.1 | XP_008763772.1 | 9 | 2127 | |||
XM_008765551.1 | XP_008763773.1 | 11 | 2119 | |||
XM_008765552.1 | XP_008763774.1 | 11 | 2100 | |||
XM_008765553.1 | XP_008763775.1 | 9 | 2019 | |||
XM_008765554.1 | XP_008763776.1 | 8 | 2073 | |||
XM_008765555.1 | XP_008763777.1 | 8 | 2058 | |||
XM_008765556.1 | XP_008763778.1 | 7 | 2001 | |||
XM_008765557.1 | XP_008763779.1 | 8 | 949 | |||
XM_008765558.1 | XP_008763780.1 | 8 | 956 | |||
XM_008765559.1 | XP_008763781.1 | 8 | 936 | |||
XM_008765560.1 | XP_008763782.1 | 8 | 934 | |||
XM_008765561.1 | XP_008763783.1 | 7 | 877 | |||
XM_008765562.1 | XP_008763784.1 | 7 | 862 | |||
GenBank mRNA | AF145050.1 | - | 2 | 129 | ||
BC079391.1 | - | 8 | 1945 | 7111852 | ||
FQ220813.1 | - | 8 | 906 | |||
FQ225933.1 | - | 8 | 941 | |||
FQ227443.1 | - | 8 | 917 | |||
FQ233760.1 | - | 8 | 897 |