Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013104.1 | NP_001013122.1 | 3 | 1273 | ||
XM_008765549.1 | XP_008763771.1 | 3 | 3337 | |||
XM_008765550.1 | XP_008763772.1 | 3 | 1398 | |||
XM_008765551.1 | XP_008763773.1 | 5 | 1390 | |||
XM_008765552.1 | XP_008763774.1 | 5 | 1371 | |||
XM_008765553.1 | XP_008763775.1 | 3 | 1290 | |||
XM_008765554.1 | XP_008763776.1 | 3 | 1401 | |||
XM_008765557.1 | XP_008763779.1 | 3 | 277 | |||
XM_008765558.1 | XP_008763780.1 | 3 | 284 | |||
XM_008765559.1 | XP_008763781.1 | 3 | 264 | |||
GenBank mRNA | BC079391.1 | - | 3 | 1273 | 7111852 | |
FQ220813.1 | - | 3 | 234 | |||
FQ225933.1 | - | 3 | 269 | |||
FQ227443.1 | - | 3 | 245 | |||
FQ233760.1 | - | 3 | 225 |