Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017649.4 | NP_060119.3 | 1 | 1380 | ||
NM_199076.2 | NP_951058.1 | 1 | 1380 | |||
NM_199077.2 | NP_951059.1 | 1 | 1380 | |||
XM_005269933.4 | XP_005269990.1 | 1 | 1405 | |||
XM_006717908.2 | XP_006717971.1 | 1 | 1405 | |||
XM_011539911.2 | XP_011538213.1 | 1 | 1405 | |||
GenBank mRNA | AF216962.1 | - | 1 | 591 | ||
AK023066.1 | - | 1 | 494 | |||
AK023479.1 | - | 1 | 1341 | |||
BC021222.2 | - | 1 | 1380 | 4308662 |