Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013877.1 | NP_001013899.1 | 7 | 1034 | ||
XM_008770455.1 | XP_008768677.1 | 7 | 1155 | |||
GenBank mRNA | BC083604.1 | - | 7 | 1034 | 7133284 |