Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013882.2 | NP_001013904.2 | 1 | 181 | ||
NM_001161512.1 | NP_001154984.1 | 1 | 397 | |||
GenBank mRNA | BC087138.1 | - | 1 | 136 | 7125992 | |
BP504069.1 | - | 399 | ||||
DY317307.1 | - | 1 | 149 | |||
EV776012.1 | - | 1 | 206 |