Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017377.1 | NP_001017377.1 | 3 | 1944 | ||
XM_006245871.2 | XP_006245933.1 | 3 | 1691 | |||
GenBank mRNA | BC092635.1 | - | 3 | 1944 | 7315721 |