Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017995.2 | NP_001017995.1 | 13 | 6264 | ||
XM_017009351.1 | XP_016864840.1 | 14 | 6348 | |||
GenBank mRNA | AB037716.1 | - | 2 | 5014 | ||
AK000838.1 | - | 1 | 119 | |||
AK095834.1 | - | 1 | 1146 | |||
BC016513.2 | - | 1 | 272 | |||
BC065230.1 | - | 1 | 32 | |||
BC110316.1 | - | 1 | 456 |