Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bles03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024233.2 | NP_001019404.2 | 2 | 1514 | ||
NM_001243534.1 | NP_001230463.1 | 2 | 1528 | |||
GenBank mRNA | AF502571.2 | - | 1 | 721 | ||
BC093387.1 | - | 2 | 1373 | 7389419 | ||
FQ233132.1 | - | 2 | 1407 |