Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024743.1 | NP_001019914.1 | 9, 10 | 1675 | ||
XM_006251538.1 | XP_006251600.1 | 9, 10 | 1627 | |||
GenBank mRNA | BC084711.1 | - | 4, 5 | 782 | ||
BC097369.1 | - | 9, 10 | 1675 | 7443949 | ||
FQ214781.1 | - | 4, 5 | 857 |