Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024750.1 | NP_001019921.1 | 12 | 1533 | ||
GenBank mRNA | BC098010.1 | - | 12 | 1533 | 7454306 | |
FQ211238.1 | - | 1534 |