Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024979.1 | NP_001020150.1 | 8 | 945 | ||
XM_008761129.1 | XP_008759351.1 | 9 | 1476 | |||
XM_008761130.1 | XP_008759352.1 | 9 | 1526 | |||
XM_008761131.1 | XP_008759353.1 | 8 | 1462 | |||
XM_008761132.1 | XP_008759354.1 | 9 | 1640 | |||
GenBank mRNA | BC093381.1 | - | 8 | 945 | 7388663 | |
FQ227539.1 | - | 1 | 248 | |||
FQ233538.1 | - | 5 | 667 |