Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024992.1 | NP_001020163.1 | 6 | 751 | ||
GenBank mRNA | BC091246.1 | - | 6 | 751 | 7366299 | |
FQ215691.1 | - | 6 | 755 | |||
FQ216010.1 | - | 6 | 775 | |||
FQ223430.1 | - | 6 | 778 |