Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172151.4 | NP_742163.4 | 7 | 980 | ||
GenBank mRNA | AY668947.1 | - | 7 | 854 | ||
AY894892.1 | - | 7 | 980 |