Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161584.1 | NP_001155056.1 | 2, 3 | 347 | ||
NM_017746.3 | NP_060216.2 | 2, 3 | 403 | |||
XM_011518798.1 | XP_011517100.1 | 2, 3 | 365 | |||
XM_017014848.1 | XP_016870337.1 | 2, 3 | 365 | |||
GenBank mRNA | AB060968.1 | - | 2, 3 | 319 | ||
AK000294.1 | - | 2, 3 | 319 | |||
AK298916.1 | - | 2, 3 | 322 | |||
AK302619.1 | - | 2, 3 | 350 | |||
AK303662.1 | - | 2, 3 | 316 | |||
AY598337.1 | - | 1, 2 | 188 | |||
BC030652.2 | - | 2, 3 | 239 | 5268105 | ||
BP336313.1 | - | 2, 3 | 403 |