Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009909.3 | NP_001009909.2 | 2 | 389 | ||
NM_001252008.1 | NP_001238937.1 | 2 | 385 | |||
NM_001252010.1 | NP_001238939.1 | 2 | 389 | |||
XM_011520056.2 | XP_011518358.1 | 2 | 3045 | |||
XM_017017648.1 | XP_016873137.1 | 2 | 302 | |||
XM_017017649.1 | XP_016873138.1 | 2 | 302 | |||
XM_017017650.1 | XP_016873139.1 | 2 | 389 | |||
XM_017017651.1 | XP_016873140.1 | 2 | 389 | |||
GenBank mRNA | AK127695.1 | - | 2 | 389 | ||
AK309077.1 | - | 2 | 164 | |||
AL832641.3 | - | 2 | 301 | |||
AY164460.1 | - | 2 | 345 | |||
AY358386.1 | - | 2 | 220 | |||
BC132829.1 | - | 2 | 181 | 40146851 | ||
BC144177.1 | - | 2 | 177 | |||
DA706662.1 | - | 2 | 389 |