Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013743.2 | NP_001013765.2 | 4 | 977 | ||
XM_006718832.3 | XP_006718895.1 | 4 | 1030 | |||
XM_011542802.2 | XP_011541104.1 | 3 | 1099 | |||
GenBank mRNA | AY634366.1 | - | 4 | 968 |