Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCAS3 SIRNA Assays ›
Gene Symbol:
BCAS3
Gene Name:
BCAS3, microtubule associated cell migration factor
Gene Aliases:
GAOB1, MAAB
Chromosome Location:
Chr.17: 60677811 - 61392838 on Build GRCh38
UniGene ID:
Hs.655028
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25371
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCAS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099432.1
NP_001092902.1
21, 22
2292
NM_001320470.1
NP_001307399.1
20, 21
2247
NM_017679.3
NP_060149.3
20, 21
2247
XM_005257472.1
XP_005257529.1
21, 22
2243
XM_011524939.1
XP_011523241.1
22, 23
2333
XM_011524940.1
XP_011523242.1
21, 22
2288
XM_011524941.1
XP_011523243.1
20, 21
2198
XM_011524942.2
XP_011523244.1
22, 23
2333
XM_011524943.2
XP_011523245.1
21, 22
2288
XM_011524944.1
XP_011523246.1
22, 23
2333
XM_011524945.2
XP_011523247.1
21, 22
2288
XM_011524946.2
XP_011523248.1
22, 23
2333
XM_011524947.1
XP_011523249.1
22, 23
2333
XM_011524948.2
XP_011523250.1
22, 23
2333
XM_011524949.2
XP_011523251.1
22, 23
2333
XM_011524950.1
XP_011523252.1
22, 23
2333
XM_011524951.2
XP_011523253.1
22, 23
2333
XM_017024783.1
XP_016880272.1
22, 23
2333
XM_017024784.1
XP_016880273.1
22, 23
2333
XM_017024785.1
XP_016880274.1
22, 23
2333
XM_017024786.1
XP_016880275.1
21, 22
2243
XM_017024787.1
XP_016880276.1
22, 23
2333
XM_017024788.1
XP_016880277.1
22, 23
2333
XM_017024789.1
XP_016880278.1
22, 23
2333
XM_017024790.1
XP_016880279.1
21, 22
2243
XM_017024791.1
XP_016880280.1
20, 21
2198
XM_017024792.1
XP_016880281.1
21, 22
2243
XM_017024793.1
XP_016880282.1
20, 21
2198
XM_017024794.1
XP_016880283.1
22, 23
2333
XM_017024795.1
XP_016880284.1
21, 22
2243
XM_017024796.1
XP_016880285.1
13, 14
1669
GenBank mRNA
AF260268.1
-
5, 6
613
AF361219.1
-
21, 22
2186
AK000135.1
-
6, 7
725
AK022526.1
-
12, 13
1496
AK023054.1
-
12, 13
1594
AK025510.1
-
21, 22
2208
AK225757.1
-
20, 21
2206
AK304791.1
-
13, 14
1521
AK314744.1
-
20, 21
2192
AL831895.1
-
7, 8
763
BC001250.1
-
6, 7
734
BC117275.1
-
21, 22
2214
40125826
BC143386.1
-
21, 22
2214
9051892
BC143387.1
-
20, 21
2169
9051893
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
transcription, DNA-templated
Golgi organization
positive regulation of endothelial cell migration
vesicle-mediated transport
microtubule organizing center organization
negative regulation of GTPase activity
tube formation
response to starvation
positive regulation of catalytic activity
positive regulation of GTPase activity
positive regulation of transcription from RNA polymerase II promoter
positive regulation of filopodium assembly
negative regulation of focal adhesion assembly
cellular response to estrogen stimulus
positive regulation of intracellular protein transport
activation of GTPase activity
regulation of establishment of cell polarity
positive regulation of actin cytoskeleton reorganization
chromatin binding
protein binding
transcription factor binding
acetyltransferase activator activity
histone acetyltransferase binding
nuclear hormone receptor binding
histone binding
beta-tubulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0