Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025278.1 | NP_001020449.1 | 3, 4 | 797 | ||
XM_008770428.1 | XP_008768650.1 | 3, 4 | 741 | |||
XM_008770429.1 | XP_008768651.1 | 3, 4 | 741 | |||
GenBank mRNA | BC093611.1 | - | 3, 4 | 797 | 7307810 |