Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSANTD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318747.1 | NP_001305676.1 | 3 | 1883 | ||
NM_001318748.1 | NP_001305677.1 | 3 | 1879 | |||
NM_001318749.1 | NP_001305678.1 | 3 | 1597 | |||
NM_001318750.1 | NP_001305679.1 | 3 | 1524 | |||
NM_032424.2 | NP_115800.1 | 3 | 2757 | |||
XM_011543022.2 | XP_011541324.1 | 4 | 2785 | |||
XM_017018416.1 | XP_016873905.1 | 4 | 1895 | |||
XM_017018417.1 | XP_016873906.1 | 3 | 1482 | |||
GenBank mRNA | AB058729.1 | - | 3 | 2697 | ||
AK027790.1 | - | 2 | 917 | |||
AL834523.1 | - | 3 | 1447 | |||
BC041118.1 | - | 3 | 1454 | 5526792 |