Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316309.1 | NP_001303238.1 | 3 | 593 | ||
XM_017013385.1 | XP_016868874.1 | 3 | 587 | |||
XM_017013386.1 | XP_016868875.1 | 3 | 587 | |||
XM_017013387.1 | XP_016868876.1 | 3 | 528 | |||
XM_017013388.1 | XP_016868877.1 | 3 | 587 | |||
XM_017013389.1 | XP_016868878.1 | 3 | 587 | |||
XM_017013390.1 | XP_016868879.1 | 3 | 587 | |||
XM_017013391.1 | XP_016868880.1 | 3 | 587 | |||
XM_017013392.1 | XP_016868881.1 | 3 | 587 | |||
XM_017013393.1 | XP_016868882.1 | 3 | 587 | |||
XM_017013394.1 | XP_016868883.1 | 2 | 373 | |||
GenBank mRNA | AB058778.1 | - | 3 | 559 |