Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ddx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053414.1 | NP_445866.1 | 19 | 1614 | ||
GenBank mRNA | BC082049.1 | - | 19 | 1614 | 7131804 | |
FQ226373.1 | - | 7 | 640 |