Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vegfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053549.1 | NP_446001.1 | 3 | 436 | ||
XM_006230910.2 | XP_006230972.1 | 3 | 533 | |||
XM_006230911.2 | XP_006230973.1 | 3 | 534 | |||
XM_006230912.2 | XP_006230974.1 | 3 | 524 | |||
GenBank mRNA | AF032925.1 | - | 1 | 32 | ||
BC076395.1 | - | 3 | 436 | 7109119 |