Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053976.1 | NP_446428.1 | 6, 7 | 1191 | ||
GenBank mRNA | BC091275.1 | - | 6, 7 | 1191 | 7383311 |