Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138370.2 | NP_612379.2 | 2, 3 | 937 | ||
GenBank mRNA | BC094697.1 | - | 2, 3 | 345 | 30529646 | |
BC110513.1 | - | 2, 3 | 374 | 40031755 | ||
BC110514.1 | - | 2, 3 | 374 | 40031756 | ||
CF131963.1 | - | 2, 3 | 433 |