Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308350.1 | NP_001295279.1 | 26 | 3628 | ||
NM_001322294.1 | NP_001309223.1 | 26 | 3631 | |||
NM_001322295.1 | NP_001309224.1 | 25 | 3544 | |||
NM_001322296.1 | NP_001309225.1 | 24 | 3442 | |||
NM_001322297.1 | NP_001309226.1 | 25 | 3506 | |||
NM_001322298.1 | NP_001309227.1 | 25 | 3605 | |||
NM_001322299.1 | NP_001309228.1 | 25 | 3632 | |||
NM_001322300.1 | NP_001309229.1 | 25 | 3525 | |||
NM_017699.2 | NP_060169.2 | 25 | 3613 | |||
NR_136280.1 | 26 | 3634 | ||||
NR_136281.1 | 25 | 3573 | ||||
NR_136282.1 | 25 | 3554 | ||||
NR_136283.1 | 24 | 3417 | ||||
NR_136284.1 | 22 | 3324 | ||||
NR_136285.1 | 21 | 3187 | ||||
XM_011512939.2 | XP_011511241.1 | 25 | 3576 | |||
XM_011512941.2 | XP_011511243.1 | 25 | 3474 | |||
XM_017006664.1 | XP_016862153.1 | 23 | 3504 | |||
XM_017006665.1 | XP_016862154.1 | 24 | 4019 | |||
XM_017006666.1 | XP_016862155.1 | 23 | 2496 | |||
XM_017006667.1 | XP_016862156.1 | 21 | 2363 | |||
XM_017006668.1 | XP_016862157.1 | 14 | 1820 | |||
GenBank mRNA | BE839090.1 | - | 1 | 52 |