Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000985.4 | NP_000976.1 | 5 | 668 | ||
NM_001035006.2 | NP_001030178.1 | 5 | 419 | |||
NM_001199340.1 | NP_001186269.1 | 6 | 517 | |||
NM_001199341.1 | NP_001186270.1 | 5 | 518 | |||
NM_001199342.1 | NP_001186271.1 | 6 | 616 | |||
NM_001199343.1 | NP_001186272.1 | 6 | 599 | |||
NM_001199344.1 | NP_001186273.1 | 5 | 501 | |||
NM_001199345.1 | NP_001186274.1 | 4 | 378 | |||
GenBank mRNA | AK304659.1 | - | 4 | 292 | ||
AK311828.1 | - | 5 | 333 | |||
BC000502.2 | - | 4 | 571 | 2821464 | ||
BC017831.1 | - | 1 | 337 | 4251433 | ||
BC066323.1 | - | 1 | 333 | 4797583 | ||
BC066324.1 | - | 1 | 337 | 4799029 | ||
BC106031.1 | - | 1 | 304 | 4540700 | ||
BF670086.1 | - | 514 | ||||
BG497914.1 | - | 5 | 585 | |||
BG758380.1 | - | 6 | 535 | |||
BG827384.1 | - | 5 | 419 | |||
BI086293.1 | - | 5 | 390 | |||
BQ877039.1 | - | 6 | 557 | |||
BQ882430.1 | - | 5 | 483 | |||
CB178432.1 | - | 4 | 584 | |||
CD386294.1 | - | 5 | 438 | |||
CD387752.1 | - | 5 | 523 | |||
X53777.1 | - | 4 | 436 |
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 4 | 311 | ||
NM_001199356.1 | NP_001186285.1 | 4 | 378 | |||
GenBank mRNA | BI461805.1 | - | 4 | 297 |