Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271420.1 | NP_001258349.1 | 3 | 956 | ||
NM_017775.3 | NP_060245.3 | 3 | 819 | |||
XM_017024801.1 | XP_016880290.1 | 3 | 732 | |||
XM_017024802.1 | XP_016880291.1 | 3 | 732 | |||
GenBank mRNA | AK000350.1 | - | 1 | 35 | ||
AK055780.1 | - | 3 | 512 | |||
AK056878.1 | - | 3 | 731 | |||
AK297783.1 | - | 3 | 375 | |||
BC011698.2 | - | 3 | 358 | |||
BC073796.1 | - | 3 | 362 | |||
BC105128.1 | - | 3 | 819 | 8144131 | ||
BC112107.1 | - | 3 | 819 | 8327575 |