Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF780B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005851.2 | NP_001005851.1 | 4 | 519 | ||
XM_005258590.4 | XP_005258647.1 | 6 | 751 | |||
XM_005258591.4 | XP_005258648.1 | 5 | 3340 | |||
XM_005258592.3 | XP_005258649.1 | 6 | 659 | |||
XM_005258593.2 | XP_005258650.1 | 6 | 735 | |||
XM_005258595.3 | XP_005258652.1 | 5 | 585 | |||
XM_006723072.3 | XP_006723135.1 | 6 | 3501 | |||
XM_011526593.2 | XP_011524895.1 | 5 | 531 | |||
XM_017026425.1 | XP_016881914.1 | 6 | 3498 | |||
XM_017026426.1 | XP_016881915.1 | 6 | 603 | |||
XM_017026427.1 | XP_016881916.1 | 5 | 582 | |||
GenBank mRNA | AK127063.1 | - | 6 | 712 | ||
BC136960.1 | - | 4 | 499 | 9020952 |