Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009703.1 | NP_001009703.1 | 4 | 461 | ||
GenBank mRNA | BC088422.1 | - | 4 | 461 | 7123664 | |
FQ219234.1 | - | 4 | 469 |