Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010895.2 | NP_001010895.1 | 14 | 2598 | ||
GenBank mRNA | AL389952.1 | - | 11 | 1453 | ||
BC035183.1 | - | 15 | 3222 | 5266310 | ||
BC075860.1 | - | 13 | 1839 | |||
BC140702.1 | - | 14 | 2341 | 9021570 | ||
BC144261.1 | - | 13 | 2147 |