Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010895.2 | NP_001010895.1 | 4 | 1134 | ||
NM_020207.4 | NP_064592.2 | 4 | 1134 | |||
XM_011518641.2 | XP_011516943.1 | 4 | 1134 | |||
XM_011518645.2 | XP_011516947.1 | 4 | 1134 | |||
XM_011518646.2 | XP_011516948.1 | 4 | 1134 | |||
XM_011518647.1 | XP_011516949.1 | 4 | 1134 | |||
XM_011518650.2 | XP_011516952.1 | 3 | 286 | |||
GenBank mRNA | AK308409.1 | - | 4 | 1066 | ||
AL389953.1 | - | 4 | 1050 | |||
BC035183.1 | - | 5 | 1758 | 5266310 | ||
BC075860.1 | - | 3 | 375 | |||
BC140702.1 | - | 4 | 877 | 9021570 |