Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012158.1 | NP_001012158.1 | 5 | 784 | ||
GenBank mRNA | AB296367.1 | - | 4 | 693 | ||
AB296368.1 | - | 3 | 549 | |||
BC087079.1 | - | 5 | 784 | 7135920 |