Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012186.2 | NP_001012186.2 | 7 | 632 | ||
XM_006234782.2 | XP_006234844.1 | 8 | 676 | |||
GenBank mRNA | BC087048.1 | - | 7 | 611 | 7112790 |