Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174081.1 | NP_001167552.1 | 2 | 254 | ||
NM_001174082.1 | NP_001167553.1 | 2 | 285 | |||
NM_017852.3 | NP_060322.1 | 2 | 285 | |||
GenBank mRNA | AB209300.1 | - | 2 | 209 | ||
AF298547.1 | - | 2 | 202 | |||
AF310106.1 | - | 2 | 230 | |||
AF464764.1 | - | 2 | 185 | |||
AK000517.1 | - | 2 | 230 | |||
AK025952.1 | - | 2 | 194 | |||
AK223253.1 | - | 2 | 194 | |||
AK223269.1 | - | 2 | 194 | |||
AK292681.1 | - | 2 | 222 | |||
BC001039.2 | - | 1 | 165 | 3504594 | ||
BC003592.1 | - | 2 | 209 | 2821428 | ||
BC039269.1 | - | 2 | 254 | 5298283 | ||
DA856893.1 | - | 2 | 254 | |||
DB046128.1 | - | 2 | 285 |