Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012212.1 | NP_001012212.1 | 8 | 1120 | ||
GenBank mRNA | BC079049.1 | - | 8 | 1120 | 7115321 |