Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC25A53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012755.4 | NP_001012773.2 | 2 | 902 | ||
XM_005262129.4 | XP_005262186.1 | 2 | 848 | |||
XM_011530952.2 | XP_011529254.1 | 3 | 1045 | |||
XM_011530953.2 | XP_011529255.1 | 3 | 876 | |||
XM_017029514.1 | XP_016885003.1 | 3 | 1687 | |||
XM_017029515.1 | XP_016885004.1 | 4 | 1195 | |||
XM_017029516.1 | XP_016885005.1 | 3 | 1818 | |||
GenBank mRNA | AL833609.1 | - | 2 | 903 | ||
BC140812.1 | - | 1 | 762 | 9021680 | ||
BC140814.1 | - | 1 | 762 | 9021682 |