Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305544.1 | NP_001292473.1 | 2 | 955 | ||
NM_017763.5 | NP_060233.3 | 2 | 1158 | |||
XM_011524955.2 | XP_011523257.1 | 2 | 1057 | |||
XM_017024800.1 | XP_016880289.1 | 3 | 2384 | |||
GenBank mRNA | AB081837.1 | - | 2 | 695 | ||
AK000322.1 | - | 1 | 375 | |||
AK291027.1 | - | 2 | 928 | |||
AK296769.1 | - | 1 | 271 | |||
BC109028.2 | - | 1 | 404 | 40027722 | ||
CR627423.1 | - | 2 | 955 | |||
CR749257.1 | - | 1 | 2163 |