Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013202.2 | NP_001013220.1 | 10 | 1544 | ||
GenBank mRNA | AB119249.1 | - | 10 | 1545 | ||
AB212889.1 | - | 10 | 1545 | |||
AB306511.1 | - | 9 | 1444 |