Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013210.1 | NP_001013228.1 | 1 | 273 | ||
GenBank mRNA | BC083746.1 | - | 1 | 273 | 7192246 | |
FQ216652.1 | - | 1 | 320 |
Gene Symbol: LOC100911857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752244.3 | XP_003752292.3 | 161 |