Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013405.2 | NP_001013423.2 | 4 | 433 | ||
GenBank mRNA | BC080690.1 | - | 4 | 392 | 30111134 |