Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC388692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111933.1 | 2 | 2208 | |||
GenBank mRNA | AK123662.1 | - | 1 | 3291 | ||
BC141801.1 | - | 1 | 2051 |
Gene Symbol: FAM231D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111934.1 | 2 | 1887 |
Gene Symbol: LOC103091866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110832.1 | - | 3 | 916 |